Order Entry
Austria
ContactUsLinkComponent
Syringe cap, MULTI-AD®
Syringe cap, MULTI-AD®
Catalog # BRAU418012
Supplier:  B. Braun
undefined
Syringe cap, MULTI-AD®
Catalog # BRAU418012
Supplier:  B. Braun

Specifications

  • Colour:
    Red
  • Description:
    Luer-Lock syringe caps
  • Seq:
    01
  • Pk:
    500

Specifications

About this item

Red syringe cap for capping Luer slip or Luer-Lock syringes.

  • Sterile
  • One-hand attachment
  • Contains PVC
  • Blister package protects of unopened caps and aids in capping

The componenets do not contain BPA, DEHP and natural rubber.

Packaging: Unique 10 per blister package protects sterility of unopened caps and aids in capping. 50 packs per case.