Cold trap, type S
About this item
Type S cold trap has a conical ground joint NS 45/40 as connection part. The condensation area can be separated from the upper section of cold trap. Therefore it is easy to pour out the condensate and to clean the whole cold trap. The connection part NS 45/40 is secured by a spring clamp out of steel.
Certifications: Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.
2 Options Available Below
Product Details & Documents
Type S cold trap has a conical ground joint NS 45/40 as connection part. The condensation area can be separated from the upper section of cold trap. Therefore it is easy to pour out the condensate and to clean the whole cold trap. The connection part NS 45/40 is secured by a spring clamp out of steel.
Certifications: Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.
Product Family Options
Product Information
- AvailabilityPrice
- KGWA17405Cold finger, S 29-Z, borosilicate glass 3,3, spherical joint S29, 150 ml condensate capacity and length of 200 mmCold finger, S 29-Z, borosilicate glass 3,3, spherical joint S29, 150 ml condensate capacity and length of 200 mmLog in for availability1 * 1 ST548,00 €
- KGWA17407Cold trap, S 29-GL-Z, borosilicate glass 3,3, with threaded glass joint GL 18, 150 ml condensate capacity and length of 200 mmCold trap, S 29-GL-Z, borosilicate glass 3,3, with threaded glass joint GL 18, 150 ml condensate capacity and length of 200 mmLog in for availability1 * 1 ST480,00 €