Order Entry
Austria
ContactUsLinkComponent
VWR® pHenomenal® OPOX 11-3 IDP Optical Dissolved Oxygen Sensor
VWR® pHenomenal® OPOX 11-3 IDP Optical Dissolved Oxygen Sensor
Catalog # 664-0182
Supplier:  VWR Collection
undefined
VWR® pHenomenal® OPOX 11-3 IDP Optical Dissolved Oxygen Sensor
Catalog # 664-0182
Supplier:  VWR Collection

Specifications

  • Connection:
    1.5 m cable, IDP-plug
  • Ø×L:
    15,3×140 mm
  • Type:
    pHenomenal® OPOX 11-3
  • Description:
    pHenomenal® OPOX 11-3 optical oxygen probe with built-in temperature sensor
  • Material:
    Epoxy
  • Pk:
    1
  • Temp.:
    0...+50 °C

Specifications

About this item

This IDP (Intelligent Digital Probe) dissolved oxygen/temperature sensor can be used with the pHenomenal® MD 8000 H and MD 8000 L multi-parameter meters.

  • Post moders type of oxygen measurement: No chemicals, no electrolytes
  • Calibration data (serial number, calibration status) is stored in the electrode/probe, transmitted to the meter and displayed
  • Proven reliability and accuracy
  • Correct measurement results due to direct signal processing in sensor and digital transmission of values
  • All parameters are easily measured with cable lengths of up to 100 metres
  • Same universal connector for all electrodes and probes

Measuring range concentration: 0,00 to 20,00 mg/L

Measuring range saturation: 0,0 to 200,0%

Measuring range partial pressure: 0,0 to 400 hPa

Accuracy: ±0,2 °C

The electrodes can provide the following data, shown on the meter's screen:
- Model name
- Serial number
- Calibration status (+++, ++, +, −, blank)
- The calibration history includes the last 10 calibrations, including buffers used, mV values.

Delivery: Includes optical DO sensor and manual only.