ContactUsLinkComponent
Slide printer, Signature®
Slide printer, Signature®
Catalog # 720-2826
undefined
Slide printer, Signature®
Catalog # 720-2826
Specifications
  • Weight:
    7,62
  • Description:
    Signature® slide printer
  • W×D×H:
    180×320×219 mm
  • Pk:
    1
Specifications
About this item
Compact, economical thermal transfer printer, from Primera Technologies, prints directly onto slides, eliminatiing handwriting or difficult to apply labels. Colour printing allows the user to use only white frosted slides and still retain colour coding/differentiation. Slides to be printed are loaded into a cartridge with capacity up to 100 slides.
  • On demand black or colour printing
  • Print text, graphics, logos, linear and 2-D barcodes
  • 300 dpi resolution
  • Chemical resistant ink
  • Reduce risk of incorrect label identification
Print speed: Up to 9 slides/minute (monochrome); 5 slides/minute (solid colours); 4 slides/minute (process colours).