Order Entry
Austria
ContactUsLinkComponent
Small-scale SEC columns, Superose 6 Increase
Small-scale SEC columns, Superose 6 Increase
Supplier:  Cytiva
Certificates
undefined
Small-scale SEC columns, Superose 6 Increase
Supplier:  Cytiva
Certificates

Specifications

  • Catalog No:
  • 29-0915-96
  • 29-0915-97
  • 29-0915-98
  • Max. pressure:
  • 30 bar
  • 30 bar
  • 30 bar
  • pH range:
  • 3 - 12
  • 3 - 12
  • 3 - 12
  • Storage:
  • 4 to 30° C, 20% ethanol
  • 4 to 30° C, 20% ethanol
  • 4 to 30° C, 20% ethanol

Specifications

Frequently Bought Together