Orders LinkContactUsLinkComponent
Forceps, blunt
Forceps, blunt
Straight, blunt, 115 mm
Catalog # BOCH1900
Supplier:   BOCHEM
Forceps, blunt
Catalog # BOCH1900
Supplier:  BOCHEM
Forceps, blunt
Forceps, blunt
Straight, blunt, 115 mm
Price
70,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Titanium
  • Ridges at the top
  • Blunt tip
Specifications
  • Type:
    Straight, blunt
  • Length:
    115 mm