Orders LinkContactUsLinkComponent
VWR®, Sampling Scoops
VWR®, Sampling Scoops
VWR®, Sampling Scoops
Supplier:  VWR Collection
Certificates
VWR®, Sampling Scoops
VWR®, Sampling Scoops
6 Options Available Below
Product Details & Documents
PS, white. These sterile scoops are ideal for sampling powder, granules and pastes.
  • Individually packed
  • Sterilised by gamma radiation
Thanks to the special handle construction, and the slightly angled edge, the scoop stands horizontally and can be laid down without the material trickling out.
Certifications: Production, assembly and packaging according to cleanroom Class 7 (10000). According to EU food and FDA guidelines.
Caution: *The product(s) below are outside the scope of the EU Directive 2019/904.
Product Family Options
Product Information
  • Capacity
    Length
    Colour
    Availability
    Price
  • 25 ml
    141 mm
    White
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 214,00 €
    214,00 €
    1 * 100 ST
       214,00 €
  • 50 ml
    161 mm
    White
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 235,00 €
    235,00 €
    1 * 100 ST
       235,00 €
  • 100 ml
    205 mm
    White
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 237,00 €
    237,00 €
    1 * 100 ST
       237,00 €
  • 150 ml
    216 mm
    White
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 251,00 €
    251,00 €
    1 * 100 ST
       251,00 €
  • 250 ml
    232 mm
    White
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 292,00 €
    292,00 €
    1 * 100 ST
       292,00 €
  • 500 ml
    280 mm
    White
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 501,00 €
    501,00 €
    1 * 100 ST
       501,00 €