Orders LinkContactUsLinkComponent
VWR®, Sampling Spoons
VWR®, Sampling Spoons
VWR®, Sampling Spoons
Supplier:  VWR Collection
Certificates
VWR®, Sampling Spoons
VWR®, Sampling Spoons
2 Options Available Below
Product Details & Documents
PS, white. These disposable spoons are produced in a cleanroom environment and sterilised by gamma radiation. They are ideal for sampling powder, granules and pastes.
  • Available in two different sizes: 2,5 ml (similar to a teaspoon) and 10 ml (similar to a tablespoon)
  • Production, assembly and packaging according to cleanroom Class 7 (10000)
  • According to EU food and FDA guidelines
  • Individually packed, sterilised by gamma radiation
Product Family Options
Product Information