Ionic strength adjustor for ammonia
Ammonia ionic strength adjustor, 500 ml bottle, 500 mL
Catalog # HACH2824349
Supplier: Hach
Jump to:
Product Details & Documents
Ammonia Ionic Strength Adjustor (ISA) buffer solution for ammonia determinations by electrode methods.
Specifications
Specifications
- Pk:500 mL
- Description:Ammonia ionic strength adjustor, 500 ml bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



