Bromocresol green-methyl red solution pH-indicator
Supplier: Hach
Certificates
About this item
:
MDL Number:MFCD00134126
Frequently Bought Together
VWR® Kjeldahl Tablets
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Bulb pipettes (one mark), BLAUBRAND®, class AS
Supplier: Brand
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
pH electrodes, SenTix® PLUS
Supplier: WTW
VWR®, Buffer, ammonia solution, pH 10 ready to use
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
VWR®, Disposable Weighing Dishes with Tabs
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
AMP (2-Amino-2-methylpropanol) 99%
Supplier: Thermo Fisher Scientific