Order Entry
Austria
ContactUsLinkComponent
1-(2-Pyridylazo)-2-naphthol (PAN) solution 0.3% indicator for nickel and cobalt
Health
Corrosive
Environment
Irritant
1-(2-Pyridylazo)-2-naphthol (PAN) solution 0.3% indicator for nickel and cobalt
Catalog # HACH2150232
Supplier:  Hach
CAS Number:  
undefined
Health
Corrosive
Environment
Irritant
1-(2-Pyridylazo)-2-naphthol (PAN) solution 0.3% indicator for nickel and cobalt
Catalog # HACH2150232
Supplier:  Hach
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440
  • Pk Info:
    50 - 100 tests

Specifications

About this item

 :
 :249.27 g/mol
UN:3082
ADR:9,III
 :GHS08
 :GHS05
 :GHS09
 :GHS07
Storage Temperature:Ambient
CAS Number:85-85-8
Formula:C₁₅H₁₁N₃O
MDL Number:MFCD00004071