Order Entry
Austria
Orders LinkContactUsLinkComponent
pH electrode storage solution

pH electrode storage solution

pH electrode storage solution, powder pillows (each pillow makes 25 ml of storage solution), 20
Catalog # HACH2657364
Supplier:   Hach
pH electrode storage solution
Catalog # HACH2657364
Supplier:  Hach
CAS Number:  
pH electrode storage solution
pH electrode storage solution

pH electrode storage solution, powder pillows (each pillow makes 25 ml of storage solution), 20

CAS Number:  
Price
71,00 €
Log in for availability
1 * 20 ST
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    20
  • Description:
    pH electrode storage solution, powder pillows (each pillow makes 25 ml of storage solution)

Specifications