Accessories for data loggers, Dickson®
Supplier: DICKSON
About this item
DicksonWare™ software (incl. USB or serial cable) is needed for setting up Dickson loggers. They enable viewing and downloading data, and resetting the logger to the factory settings.
2 Options Available Below
Product Details & Documents
DicksonWare™ software (incl. USB or serial cable) is needed for setting up Dickson loggers. They enable viewing and downloading data, and resetting the logger to the factory settings.
Specifications for Product Family
Specifications
- Catalog No:
- DICKA015
- 620-1515
- Interface:
- USB