
Ammonium acetate ≥99%, HiPerSolv CHROMANORM® for LC-MS
:631-61-8
:
:H₃CCOONH₄
:MFCD00013066
:77.08 g/mol
:13,00495
:114 °C
:1.17 g/cm³ (20 °C)
:136 °C
:211-162-9
:GHS07
:Ambient
- Assay (on anhydrous substance):Min. 99 %
- Ignition residue:Max. 0.01 %
- Water:Max. 2 %
- Cl (Chloride):Max. 5 ppm
- SO4 (Sulphate):Max. 10 ppm
- NO3 (Nitrate):Max. 10 ppm
- Ca (Calcium) (0.1 %):Max. 0.2 ppm
- K (Potassium) (0.1 %):Max. 0.2 ppm
- Mg (Magnesium) (0.1 %):Max. 0.2 ppm
- Na (Sodium) (0.1 %):Max. 0.5 ppm
- Fe (Iron) (0.1%):Max. 0.2 ppm
- Transmittance (210 nm) (0.1 %):Min. 10 %
- Transmittance (220 nm) (0.1 %):Min. 50 %
- Transmittance (230 nm) (0.1 %):Min. 80 %
- Transmittance (235 nm) (0.1 %):Min. 95 %
- Transmittance (245 nm) (0.1 %):Min. 99 %
- Suitable for LC-MS:Passes test
spec_VWRC84885.180 spec_VWRC84885.260
Product specification
VWR® LC-MS Solvents, Additives, Mixes and Associated Products
Product Literature
Frequently Bought Together