Orders LinkContactUsLinkComponent
Microscope slides
Microscope slides
Microscope slides
These slides are made of glass with ground edges.
Product Family Options
  • Description
    Thickness
    L×W
  • Microscope slides
    1,0 - 1,2 mm
    76×26 mm
    1 * 50 ST
    1 * 50 ST - 9,40 €
    9,40 €
    1 * 50 ST
       9,40 €
  • Microscope slides with one end frosted on both sides
    1,0 - 1,2 mm
    76×26 mm
    1 * 50 ST
    1 * 50 ST