Visual colour kit, detergents
Catalog # HACH143203
Supplier: Hach
CAS Number:
Specifications
- Chemical method:Toluidine blue-O chloroform
- Pk:1 KIT
- Description:Detergents, model DE-2
- Range:0 - 1,0 mg/l
Specifications
About this item
It is a simple, economical colour disc kit to determine the presence of detergents in surface or ground waters which indicates industrial or domestic pollution. In natural waters anionic LAS and ABS detergents cause excessive foaming, promote growth of disease producing organisms and tend to kill aquatic life. This kit achieves fast results with an portable test kit.
Delivery: Kit includes four reagents, one thimble, two test tubes, one rubber bulb, two glass tubes, glass wool, one dropper, two stoppers, a colour comparator box, one colour disc, instructions, and a carrying case.