VWR®, ICP multi-element solution
ICP multi element solution according USP 232 Oral dose 8 elements in HNO₃ 7%, Plastic bottle, 100 mL
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
ICP solution of eight elements in 7% nitric acid.
Cd 25 mg/L, Pb 5 mg/L, As 1,5 mg/L, Hg 15 mg/L, Mo 100 mg/L, Ni 500 mg/L, V 100 mg/L, Cu 1000 mg/L.
Certifications: USP 232 Oral dose
Traceable to SRM from NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Documents
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Description:ICP multi element solution according USP 232 Oral dose 8 elements in HNO₃ 7%



