ContactUsLinkComponent
VWR® ARISTAR®, ICP Multi-Element Solution, 4 Elements in Nitric Acid 7%
Catalog # 85035.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, ICP Multi-Element Solution, 4 Elements in Nitric Acid 7%
Catalog # 85035.180
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    ICP multi element solution
Specifications
About this item
ICP multi-element solution according to USP 232 dietary supplements.
Cd 5 mg/L, Pb 10 mg/L, As 15 mg/L, Hg 15 mg/L in HNO₃ 7%.
Certifications: Traceable to SRM from NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.