Culture tubes, reusable, with screw cap, KIMAX®, KIMBLE®
Supplier: DWK Life Sciences
Certificates
About this item
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
- Very good chemical resistance
- Very good temperature resistance
- Uniform wall thickness
Certifications: Tubes with PTFE-faced rubber-lined caps are designed from ASTM Specification E982, Type VI, Class B requirements.
Ordering information: Cap is supplied unattached.
Frequently Bought Together