Order Entry
Austria
ContactUsLinkComponent
Coating testers, PosiTector® 200
Coating testers, PosiTector® 200
Supplier:  Industrial Physics
Certificates
undefined
Coating testers, PosiTector® 200
Supplier:  Industrial Physics

Specifications

  • Catalog No:
  • SIMELD0214
  • SIMELD0212
  • SIMELD0213
  • SIMELD0215
  • SIMELD0210
  • SIMELD0211
  • Model:
  • 200 D1 Standard
  • 200 C1 Standard
  • 200 C3 Advanced
  • 200 D3 Advanced
  • 200 B1 Standard
  • 200 B3 Advanced
  • Accuracy:
  • ±2 µm +3 %
  • ±2 µm +3 %
  • ±2 µm +3 %
  • ±2 µm +3 %
  • ±2 µm +3 %
  • ±2 µm +3 %
  • Results stored:
  • 250
  • 250
  • 100 000
  • 100 000
  • 250
  • 100 000
  • Thickness:
  • 50 - 7600 µm
  • 50 - 3800 µm
  • 50 - 3800 µm
  • 50 - 7600 µm
  • 13 - 1000 µm
  • 13 - 1000 µm
  • W×D×H:
  • 61×28×135 mm
  • 61×28×135 mm
  • 61×28×135 mm
  • 61×28×135 mm
  • 61×28×135 mm
  • 61×28×135 mm
  • Weight:
  • 140 g
  • 140 g
  • 140 g
  • 140 g
  • 140 g
  • 140 g

Specifications

Related Information

Frequently Bought Together