Orders LinkContactUsLinkComponent
Colorimetric test kit, sulphite, TESTOVAL®
Colorimetric test kit, sulphite, TESTOVAL®
Colorimetric test kit, sulphite, TESTOVAL®
Certificates
Colorimetric test kit, sulphite, TESTOVAL®
Colorimetric test kit, sulphite, TESTOVAL®
2 Options Available Below
Product Details & Documents
Visual colour test method using a comparator disc. Two reagents.
Product Family Options
Product Information
  • Range
    Pk
    Availability
    Price
  • HEYL410634
    Sulphite, colour comparison kit
    Sulphite, colour comparison kit
    0,5-1-2,5-5-10-20 mg/l
    150 Tests
    Log in for availability
    Log in for availability
    1 * 150 Tests
    1 * 150 Tests - 146,00 €
    146,00 €
    1 * 150 Tests
       146,00 €
  • HEYL410635
    Sulphite, refill pack
    Sulphite, refill pack
    0,5-1-2,5-5-10-20 mg/l
    150 Tests
    Log in for availability
    Log in for availability
    1 * 150 Tests
    1 * 150 Tests - 97,60 €
    97,60 €
    1 * 150 Tests
       97,60 €
Accessories
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Replacement cuvettes for colour comparison kits
    Replacement cuvettes for colour comparison kits
    HEYL410001
    1
    Log in for availability
    1 * 1 ST - 12,70 €
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Replacement colour comparator, complete
    Replacement colour comparator, complete
    HEYL410636
    1
    Log in for availability
    1 * 1 ST - 77,10 €