Orders LinkContactUsLinkComponent
Evaporating flasks
Evaporating flasks
Supplier:   BUCHI
Evaporating flasks
Supplier:  BUCHI
Certificates
Evaporating flasks
Evaporating flasks
27 Options Available Below
Product Details & Documents
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For Rotavapor® rotary evaporators. Centre neck. Pear-shaped with ground joint socket or EL-flange.
Product Family Options
Flasks with standard ground joint socket
Product Information
  • Capacity
    Socket size
    Type
    Availability
    Price
  • 50 ml
    24/29 NS
    uncoated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 87,00 €
    87,00 €
    1 * 1 ST
       87,00 €
  • 50 ml
    24/40 NS
    uncoated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 80,00 €
    80,00 €
    1 * 1 ST
       80,00 €
  • 50 ml
    29/42 NS
    uncoated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 129,00 €
    129,00 €
    1 * 1 ST
       129,00 €
  • 100 ml
    24/29 NS
    uncoated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 87,00 €
    87,00 €
    1 * 1 ST
       87,00 €
  • 100 ml
    24/40 NS
    uncoated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 80,00 €
    80,00 €
    1 * 1 ST
       80,00 €
  • 100 ml
    29/42 NS
    uncoated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 144,00 €
    144,00 €
    1 * 1 ST
       144,00 €
  • 250 ml
    24/29 NS
    uncoated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 87,00 €
    87,00 €
    1 * 1 ST
       87,00 €
  • 250 ml
    24/40 NS
    uncoated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 87,00 €
    87,00 €
    1 * 1 ST
       87,00 €
  • 250 ml
    29/42 NS
    uncoated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 87,00 €
    87,00 €
    1 * 1 ST
       87,00 €
  • 500 ml
    24/40 NS
    uncoated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 80,00 €
    80,00 €
    1 * 1 ST
       80,00 €
  • 500 ml
    24/40 NS
    coated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 129,00 €
    129,00 €
    1 * 1 ST
       129,00 €
  • 500 ml
    29/42 NS
    uncoated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 80,00 €
    80,00 €
    1 * 1 ST
       80,00 €
  • 1000 ml
    24/29 NS
    uncoated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 80,00 €
    80,00 €
    1 * 1 ST
       80,00 €
  • 1000 ml
    24/40 NS
    uncoated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 80,00 €
    80,00 €
    1 * 1 ST
       80,00 €
  • 1000 ml
    24/40 NS
    coated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 125,00 €
    125,00 €
    1 * 1 ST
       125,00 €
  • 1000 ml
    29/42 NS
    uncoated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 80,00 €
    80,00 €
    1 * 1 ST
       80,00 €
  • 1000 ml
    29/42 NS
    coated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 188,00 €
    188,00 €
    1 * 1 ST
       188,00 €
  • 2000 ml
    24/29 NS
    uncoated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 129,00 €
    129,00 €
    1 * 1 ST
       129,00 €
  • 2000 ml
    24/40 NS
    uncoated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 129,00 €
    129,00 €
    1 * 1 ST
       129,00 €
  • 2000 ml
    24/40 NS
    coated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 187,00 €
    187,00 €
    1 * 1 ST
       187,00 €
  • 3000 ml
    29/32 NS
    coated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 347,00 €
    347,00 €
    1 * 1 ST
       347,00 €
  • 3000 ml
    24/40 NS
    uncoated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 175,00 €
    175,00 €
    1 * 1 ST
       175,00 €
  • 3000 ml
    24/40 NS
    coated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 231,00 €
    231,00 €
    1 * 1 ST
       231,00 €
  • 3000 ml
    29/42 NS
    uncoated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 248,00 €
    248,00 €
    1 * 1 ST
       248,00 €
  • 3000 ml
    29/42 NS
    coated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 308,00 €
    308,00 €
    1 * 1 ST
       308,00 €
  • 4000 ml
    24/40 NS
    uncoated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 276,00 €
    276,00 €
    1 * 1 ST
       276,00 €
  • 4000 ml
    24/40 NS
    coated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 327,00 €
    327,00 €
    1 * 1 ST
       327,00 €