ContactUsLinkComponent
Goggles, uvex flex seal®
Goggles, uvex flex seal®
Catalog # DALLS3405X
Supplier:  HONEYWELL SAFETY
undefined
Goggles, uvex flex seal®
Catalog # DALLS3405X
Supplier:  HONEYWELL SAFETY
Specifications
  • Type:
    Anti-fog, with fabric headband
  • Colour:
    Dark blue
  • Seq:
    02
  • Environmentally Preferable:
  • Lenses:
    Clear PC
  • Pk:
    1
Specifications
About this item
Lightweight, low-profile goggles with a silicone body and an optically correct clear polycarbonate lens provide a full field of vision protect the face against impacts and chemical splashes.
  • Cured silicone body stays soft and supple in extreme temperatures
  • Headband available in fabric or neoprene for chemical applications
  • Uvextreme® antifog lens coating and indirect venting system minimises fogging
  • Fit over most prescription eyewear
  • Economical lens replacement
Certifications: ANSI Z87+ and CSA Z94.3 standards