ContactUsLinkComponent
Anti-vibration pad, Richter
Anti-vibration pad, Richter
Catalog # DIVEVIBE-1000
Anti-vibration pad, Richter
Catalog # DIVEVIBE-1000
Specifications
  • Description:
    Anti-vibration pad
  • L×W:
    215x170 mm mm
  • Pk:
    1
Specifications
About this item
Richter anti-vibration pad dampens vibrations generated from lab equipment. It creates a stable surface that will keep equipment in place.
  • Small, lightweight design makes transportation between benches and laboratories easy
  • Pads can be stacked for heavier units or laid side-by-side for use with larger equipment
Ideal for use with vortexes, shakers, rockers and microscopes.