Order Entry
Austria
ContactUsLinkComponent
Hoechst wax C, micropowder tableting aid for X-ray analysis, Supelco®
Hoechst wax C, micropowder tableting aid for X-ray analysis, Supelco®
Catalog # 1.09014.1000
Supplier:  Merck
CAS Number:  
Hoechst wax C, micropowder tableting aid for X-ray analysis, Supelco®
Catalog # 1.09014.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Bi (Bismuth):
    ≤ 10 µg/g
  • Co (Cobalt):
    ≤ 5 µg/g
  • Cr (Chromium):
    ≤ 5 µg/g
  • Cu (Copper):
    ≤ 5 µg/g
  • Fe (Iron):
    ≤ 5 µg/g
  • Mn (Manganese):
    ≤ 5 µg/g
  • Ni (Nickel):
    ≤ 5 µg/g
  • Pb (Lead):
    ≤ 10 µg/g
  • Sr (Strontium):
    ≤ 5 µg/g
  • Tl (Thallium):
    ≤ 10 µg/g
  • Zn (Zinc):
    ≤ 5 µg/g

Specification Test Results

About this item

 :
 :593.04 g/mol
Melting Pt:144 °C
 :M-Clarity™ quality level = MQ100
Flash Pt:270 °C (DIN 51758)
Storage Temperature:Ambient
EINECS:203-755-6
CAS Number:110-30-5
Formula:C₃₈H₇₆N₂O₂
MDL Number:MFCD00059224