

Ethidium bromide 10 mg/ml in aqueous solution, Bioreagent for molecular biology, Sigma-Aldrich®
Catalog # SIALE1510-10ML
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mL
- Seq:2275
Specifications
About this item
:
ADR:6.1,III
:394.31 g/mol
UN:2810
:GHS06
:GHS08
CAS Number:1239-45-8
Formula:C₂₁H₂₀BrN₃
MDL Number:MFCD00011724