J.T.Baker® DILUT-IT™, Potassium Phosphate, Dissolution Media Concentrate
Supplier: Avantor
Certificates
About this item
DILUT-IT is a ready-to-use dissolution media concentrate, which is pre-measured and only need to be diluted with purified water.
DILUT-IT™ dissolution media concentrates are produced from raw materials in accordance with USP/EP/BP/JP guidelines under validated processes in Avantor Performance Materials' ISO certified manufacturing facilities
Frequently Bought Together