Order Entry
Austria
ContactUsLinkComponent
Bubble point test apparatus, Nalgene®
Bubble point test apparatus, Nalgene®
Catalog # NALGDS0405-0050
   Low Manufacturing Impact
undefined
Bubble point test apparatus, Nalgene®
Catalog # NALGDS0405-0050
   Low Manufacturing Impact

Specifications

  • Description:
    For MF75 filter units and bottle top filters, 50, 75 and 90 mm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    1

Specifications

About this item

This Bubble point test apparatus is used to ensure product integrity of MF75 Series filter unit or bottle top filters. Tests filter unit membranes without removing the membrane from the filter unit.

  • Cast aluminium
  • Accepts 6 mm int.Ø tubing
Delivery: Adapters and O-rings included.