Orders LinkContactUsLinkComponent
Digital timer
Digital timer
Digital dual timer for Buchholz indentation test
Catalog # SIMEDI0085
Supplier:   Industrial Physics
Digital timer
Catalog # SIMEDI0085
Supplier:  Industrial Physics
Digital timer
Digital timer
Digital dual timer for Buchholz indentation test
Price
57,90 €
Log in for availability
1 * 1 ST
Product Details & Documents
This dual digital timer can be pre-set for both 'in position' and 'recovery' time.
Certifications: Compliant with the ISO 2815-2003 standard.
Documents
Specifications
  • Description:
    Digital dual timer for Buchholz indentation test