ContactUsLinkComponent
Trolley for gas cylinders
Trolley for gas cylinders
  MSGA13267
 :  asecos
undefined
Trolley for gas cylinders
  MSGA13267
 :  asecos
  • Description:
    Trolley for carrying gas cylinders up to 4 (Ø 230 mm)
  • Pk:
    1
This trolley made of PE can equip up to 4 cylinders with a load capacity of 300 kg. Safe transport is ensured with the provision for forklift.
 : With lashing straps and safety chain.