ContactUsLinkComponent
Cold traps, BOLA
Cold traps, BOLA
Catalog # BOHLB317-60
Supplier:  Bohlender
undefined
Cold traps, BOLA
Catalog # BOHLB317-60
Supplier:  Bohlender
Specifications
  • Description:
    Cold traps, PFA, collection capacity 320 ml, O.Ø×H 60×400 mm
  • Pk:
    1
Specifications
About this item
PTFE head with two threaded necks GL14 for connection of hard-walled tubing by means of separately available laboratory screw joints. Connecting nut is made of PTFE reinforced with glassfibres. GL hose connectors are recommend for soft, flexible tubing. Temperature resistant from −200 to +250 °C with suitable vaccum transparent. PFA, transparent.