Order Entry
Austria
ContactUsLinkComponent
Solid phase extraction cartridge, Sep-Pak® XPoSure™ Plus
Solid phase extraction cartridge, Sep-Pak® XPoSure™ Plus
  WATEWAT047205
 :  WATERS
Solid phase extraction cartridge, Sep-Pak® XPoSure™ Plus
  WATEWAT047205
 :  WATERS

 

  • Particle Size:
    500 - 1000 µm
  • Pore size:
    125 Å
  • Capacity:
    0,7 ml
  • Description:
    Sep-Pak® XPoSure™ Plus short cartridge
  • Bed weight:
    350 mg
  • Pk:
    20

 

 

Specially designed for monitoring aldehydes in indoor air.