Order Entry
Austria
ContactUsLinkComponent
Reagent reservoirs, WHEATON®
Reagent reservoirs, WHEATON®
Catalog # WHEAW851368
Supplier:  DWK Life Sciences
undefined
Reagent reservoirs, WHEATON®
Catalog # WHEAW851368
Supplier:  DWK Life Sciences

Specifications

  • Packed:
    10/pack
  • Sterile:
    +
  • Type:
    12-channel, no lid
  • Capacity:
    48 ml
  • W×D×H:
    83×126×13 mm
  • Pk:
    10

Specifications

About this item

White PS, sterile. All-purpose reagent reservoirs for single and multi channel pipetting.