1-Propanol ≥99.0% (by GC, corrected for water content), BAKER ANALYZED®, J.T.Baker®
: Avantor
:71-23-8
:
:H₃CCH₂CH₂OH
:MFCD00002941
:60.1 g/mol
:13,07934
:97 °C (1013 hPa)
:–127 °C
:0.8053 g/cm³ (20 °C)
:15 °C (closed cup)
:Ambient
:200-746-9
:1274
:3,II
:GHS02
:GHS05
:GHS07
- For Laboratory,Research,or Manufacturing Use:= 99.0 %
- Assay (CH₃CH₂CH₂OH) (by GC, corrected for water):≥99.0 %
- Residue after Evaporation:≤0.0100 %
Frequently Bought Together