Triethylamine ≥99% (by GC), BAKER ANALYZED®, J.T.Baker®
Catalog # W635-07
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99 %
- Assay ((C₂H₅)₃N) (by GC, corrected for water):≥ 99 %
- Color (APHA):≤ 15
- Water (H₂O):≤ 1 %
- Identification (by IR):Passes Test
Specification Test Results
About this item
Synonyms: (Diethylamino)ethane, TEA
:
:101.19 g/mol
Flash Pt:–11 °C
EINECS:204-469-4
UN:1296
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:121-44-8
Formula:(C₂H₅)₃N
MDL Number:MFCD00009051
Boiling Pt:90
Melting Pt:–115 °C
Density:0.729 g/cm³ (20 °C)
ADR:3 II
Merck Index:13,09740
:Management of Change (MOC) category = N