
L(+)-Tartaric acid ≥99.0% (by acidimetry), crystals, BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
:87-69-4
:
:C₄H₆O₆
:MFCD00064207
:13,09158
:399 °C (1013 hPa)
:168…170 °C
:1.759 g/cm³ (20 °C)
:150 °C (closed cup)
:150.09 g/mol
:Ambient
:201-766-0
:01-2119537204-47
:GHS05
- Meets ACS Reagent Chemical Requirements:≥99.0%
- For Laboratory,Research,or Manufacturing Use:≤0.005%
- Assay (HOCO(CHOH)₂COOH) (by acidimetry):≥99.0 %
- Insoluble Matter:≤0.005 %
- Residue after Ignition:≤0.020 %
- Chloride (Cl):≤0.001 %
- Oxalate (C₂O₄):Passes Test
- Phosphate (PO₄):≤0.001 %
- Sulfur Compounds (as S):≤0.002 %
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
Frequently Bought Together