Order Entry
Austria
ContactUsLinkComponent
D-(+)-Sucrose, crystals, AR® ACS, Macron Fine Chemicals™
D-(+)-Sucrose, crystals, AR® ACS, Macron Fine Chemicals™
Certificates
undefined
D-(+)-Sucrose, crystals, AR® ACS, Macron Fine Chemicals™
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • Solubility:
    Passes Test
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Chloride (Cl):
    ≤0.005 %
  • ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • ACS - Iron (Fe):
    ≤5 ppm
  • ACS - Titrable Acid (meq/g):
    ≤0.0008
  • ACS - Invert Sugar:
    ≤0.05 %
  • ACS - Loss on Drying at 105°C:
    ≤0.03 %
  • ACS - Residue after Ignition:
    ≤0.01 %
  • ACS - Sulfate and Sulfite (as SO₄):
    ≤0.005 %
  • ACS - Specific Rotation [Å]²⁵^D (+):
    66.3 - 66.8 Degree

Specification Test Results

Related Information

Frequently Bought Together