Order Entry
Austria
ContactUsLinkComponent
Disodium hydrogen phosphate dodecahydrate 98.0-102.0%, crystals, BAKER ANALYZED®, J.T.Baker®
Disodium hydrogen phosphate dodecahydrate 98.0-102.0%, crystals, BAKER ANALYZED®, J.T.Baker®
Catalog # 3822-07
Supplier:  Avantor
CAS Number:  
undefined
Disodium hydrogen phosphate dodecahydrate 98.0-102.0%, crystals, BAKER ANALYZED®, J.T.Baker®
Catalog # 3822-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE pail, screw top
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    98.0 - 102.0%
  • Assay:
    98.0 - 102.0 %
  • Insoluble Matter:
    ≤0.005 %
  • pH of 5% Solution at 25°C:
    8.7 - 9.3
  • Chloride (Cl):
    ≤0.001 %
  • Sulfate (SO₄):
    ≤0.002 %
  • Heavy Metals (as Pb):
    ≤0.001 %
  • Trace Impurities - Iron (Fe):
    ≤0.001 %
  • Nitrogen Compounds (as N):
    ≤5 ppm

Specification Test Results

About this item

 :
 :358.14 g/mol
Merck Index:13,08733
Melting Pt:35 °C
Density:1.52 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-448-7
CAS Number:10039-32-4
Formula:Na₂HPO₄·12H₂O
MDL Number:MFCD00149181