Order Entry
Austria
ContactUsLinkComponent
Nitric acid 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Corrosive
Nitric acid 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # 5600-02
Supplier:  Avantor
CAS Number:  
undefined
Corrosive
Nitric acid 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # 5600-02
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    0.0995 - 0.1005
  • Normality:
    0.0995 - 0.1005

Specification Test Results

About this item

 :
 :63.01 g/mol
Storage Temperature:Ambient
EINECS:231-714-2
UN:3264
ADR:8,III
 :GHS05
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349