Express Eingabe
Austria
ContactUsLinkComponent
Single wafer shippers, for 5" (125 mm) wafer handling, H 22 series
Single wafer shippers, for 5" (125 mm) wafer handling, H 22 series
  216-2354
 :  ENTEGRIS
undefined
Single wafer shippers, for 5" (125 mm) wafer handling, H 22 series
  216-2354
 :  ENTEGRIS

 

  • Bezeichnung:
    Spring, PP, natural
  • Seq:
    03
  • VE:
    100

 

 

Conical pocket holds one wafer face down contacting only the edge of the wafer.

  • Spring retains wafer and minimises particle generation
  • Stackable for efficient use of storage space
  • Loose or lock stacking, either tray on tray or cover on tray
  • Convenient screw on covers
  • Large area on cover provides space for labelling