Order Entry
Austria
ContactUsLinkComponent
Homogeniser, HPH 2000/04
Homogeniser, HPH 2000/04
Supplier:  IKA
Certificates
undefined
Homogeniser, HPH 2000/04
Supplier:  IKA

Specifications

  • Catalog No:
  • IKAAU071735
  • IKAAU066126
  • IKAAU068906
  • IKAAT061555
  • Model:
  • HPH 2000/04-DH5
  • HPH 2000/04-DH5
  • HPH 2000/04-SH5
  • HPH 2000/04-SH5
  • Volume H₂O:
  • 6000 ml
  • 6000 ml
  • 6000 ml
  • 6000 ml
  • Speed range:
  • 344 min⁻¹ (fixed)
  • 344 min⁻¹ (fixed)
  • 344 min⁻¹ (fixed)
  • 344 min⁻¹ (fixed)
  • Speed adjustment:
  • fixed
  • fixed
  • fixed
  • fixed
  • Motor rating:
  • 1500 W
  • 1500 W
  • 1500 W
  • 1500 W
  • W×D×H:
  • 284×656×568 mm
  • 284×656×568 mm
  • 286×639×509 mm
  • 286×639×509 mm
  • Weight:
  • 36 kg
  • 36 kg
  • 36 kg
  • 36 kg

Specifications

Frequently Bought Together