ContactUsLinkComponent
Adhesion slides, HistoBond®
Adhesion slides, HistoBond®
Catalog # MARI0810701
Supplier:  MARIENFELD
Adhesion slides, HistoBond®
Catalog # MARI0810701
Supplier:  MARIENFELD
Specifications
  • Colour:
    White
  • Description:
    Slides with frosted ends
  • Pk:
    2.000
Specifications
About this item
HistoBond slides combine frosted ends with printed in various colours.
  • Soda lime glass of hydrolytic class 3
  • Different colours for colour coding of methods of analysis
  • Thin-coated marking area facilitates the processing on automated systems
Certifications: IVD directive 98/79/EC