Order Entry
Austria
ContactUsLinkComponent
Hydrochloric acid 0.01 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Corrosive
Irritant
Hydrochloric acid 0.01 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # 5611-02
Supplier:  Avantor
CAS Number:  
undefined
Corrosive
Irritant
Hydrochloric acid 0.01 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # 5611-02
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE bottle, narrow mouth
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    0.0095 - 0.0105
  • Normality:
    0.0095 - 0.0105
  • Sulfate (SO₄):
    ≤ 1 ppm
  • Ammonium (NH₄):
    ≤ 3 ppm
  • Trace Impurities - Heavy Metals:
    ≤ 0.5 ppm
  • Trace Impurities - Iron (Fe):
    ≤ 0.1 ppm
  • Appearance:
    Passes Test

Specification Test Results

About this item

 :
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8,III
 :Management of Change (MOC) category = NI
 :36.46 g/mol
 :GHS05
 :GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324