Order Entry
Austria
ContactUsLinkComponent
 
 
 
Hydrochloric acid concentrate for dilution to 0.01 mol (0.01N), DILUT-IT® dissolution media concentrate, J.T.Baker®
 :  Avantor
 
undefined
 
 
Hydrochloric acid concentrate for dilution to 0.01 mol (0.01N), DILUT-IT® dissolution media concentrate, J.T.Baker®
 :  Avantor
 

 

  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • STORE AT ROOM TEMPERATURE:
    For Information Only
  • Appearance:
    Passes Test
  • Normality at 26x dilution:
    0.0098 - 0.0102

 

Frequently Bought Together