Order Entry
Austria
ContactUsLinkComponent
Glycerine, anhydrous ≥99.5% (by GC), BAKER ANALYZED® ACS, J.T.Baker®
Glycerine, anhydrous ≥99.5% (by GC), BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Glycerine, anhydrous ≥99.5% (by GC), BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating)

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    Min. 99.5 %
  • For Laboratory,Research,or Manufacturing Use:
    Max. 10
  • Assay (HOCH₂CHOHCH₂OH) (by GC, corrected for water):
    ≥99.5 %
  • Color (APHA):
    ≤10
  • Residue after Ignition:
    ≤0.005 %
  • Neutrality:
    Passes Test
  • Chlorinated Compounds (as Cl):
    ≤0.003 %
  • Sulfate (SO₄):
    ≤0.001 %
  • Acrolein and Glucose:
    Passes Test
  • Fatty Acid Esters (as Butyric Acid):
    ≤0.05 %
  • Substances Darkened by H₂SO₄:
    Passes Test
  • Water (H₂O)(by Karl Fischer titrn):
    ≤0.5 %
  • Heavy Metals (as Pb):
    ≤2 ppm

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together