Pig Native Gelatin (from Skin)
Catalog # RC-052
Supplier: G-Biosciences
CAS Number:
Specifications
- Pack type:bottle
- Pk:100 g
- CAS No.:9000-70-8
- Protein/peptide type:Native
- Source:Skin
- Species:Pig
- Seq:01
- Storage conditions:Ambient
- Protein synonyms:Gelatine
- Protein/peptide name:Gelatin
- Grade:Research Grade