About this item
These aerosol-resistant tips are designed for use with RAININ Pipet-Lite, Pipet-Plus and EDP3-Plus pipettes with LTS LiteTouch Tip Ejection System. 1200 µl tips are specifically designed for use with RAININ E12-1200 and E8-1200 multi-channel pipettes. The unique design prevents drop formation and eliminates the need for touch-off between dispenses.
- Virgin PP with PE filter, sterile
- Filters are an extremely strong chemically inert matrix molded from pure virgin PE that does not flake or shed particles
- Hydrophobic filter forms a barrier against aerosols and liquids, eliminating potential sample and pipette cross contamination
- Certified free of RNase, DNase, DNA, Pyrogens, and ATP
- Extended length tips (102 mm long) will reach into the bottom of blocks, tall vials, flasks, 100 mm tubes etc.
- Low retention tips feature an ultra-hydrophobic tip surface that minimises liquid retention; ideal for pipetting proteins, enzymes and DNA
The tips are available in different packaging options:
Racks with removable cover
- Sealed bottoms and overlapping lids protect tips from dust, dirt and micro-organisms
- Two-position lid allows autoclaving with the lid in vent position
- Sturdy shelf construction allows easy tip loading with single and multi-channel pipettes
Green-Pak® Refills
- Environmentally friendly refills for racks; less plastic waste
- Refills contain 96 tips in 8×12 format, sealed in a shell of Code 1 PET recyclable plastic
- Each refill is completely sealed to protect the tips; hands and fingers never contact the tips during loading
StableRak™
- 192 tips in one double-96 rack
- Each StableRak™ contains 192 tips in one rack instead of two