Immobilised avidin for biotin purification
Supplier: G-Biosciences
Certificates
About this item
This avidin resin is designed for the single step small and large scale affinity purification of proteins and antibodies with a biotin tag. The resin can also be used for immunoprecipitations using biotin labelled antibodies.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together