Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Electrolytes
Electrode Filling Solutions
VWR®, Electrode Filling and Storage Solution
VWR®, Electrode Filling and Storage Solution
Catalog # 662-1116
Supplier:
VWR Collection
CAS Number:
SDS
Expand image
VWR®, Electrode Filling and Storage Solution
Catalog # 662-1116
Supplier:
VWR Collection
CAS Number:
SDS
Specifications
Pk:
114 mL
Description:
Calomel-free pH electrode refill solution containing potassium nitrate
Specifications
About this item
Product Details
Documentation
Product Details
Documentation