Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Environmental Analysis Standards
Standards for portable colorimeters, CAL Check™
Standards for portable colorimeters, CAL Check™
Standards for portable colorimeters, CAL Check™
Catalog # HANNHS96710-11
Supplier:
Hanna
CAS Number:
Expand image
Standards for portable colorimeters, CAL Check™
Catalog # HANNHS96710-11
Supplier:
Hanna
CAS Number:
Specifications
Pk:
1
Description:
CAL Check™ Standard for pH/free Chlorine/total Chlorine
Specifications
About this item
Product Details
Product Details